4-Chloro-5-methylpyrimidin-2-amine 98%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-4-chloro-5-methylpyrimidine

A884186-1G A884186-250MG A884186-25G A884186-5G
77036-794EA 53.05 USD
77036-794 77036-796 77036-798 77036-800
4-Chloro-5-methylpyrimidin-2-amine 98%
4-Chloro-5-methylpyrimidin-2-amine

Precaución: for Research Use Only

Formula: C₅H₆ClN₃
MW: 143.58 g/mol
MDL Number: MFCD00725898
CAS Number: 20090-58-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR