GSK-2801 98+%

Supplier: Ambeed
Warning

Synonyms: GSK2801, 1-(1-(2-(Methylsulphonyl)phenyl)-7-propoxyindolizin-3-yl)ethanone

A884058-10MG A884058-1MG A884058-50MG A884058-5MG
76993-752EA 35.01 USD
76993-752 76993-754 76993-756 76993-758
GSK-2801 98+%
GSK-2801

Precaución: for Research Use Only

Formula: C₂₀H₂₁NO₄S
MW: 371.54 g/mol
MDL Number: MFCD26142953
CAS Number: 1619994-68-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR