tert-Butyl ((3-bromopyridin-4-yl)methyl)carbamate 97%

Supplier: Ambeed
Warning

A884004-100MG A884004-5G
76993-748EA 80.63 USD
76993-748 76993-750
tert-Butyl ((3-bromopyridin-4-yl)methyl)carbamate 97%
tert-Butyl ((3-bromopyridin-4-yl)methyl)carbamate

Precaución: for Research Use Only

Formula: C₁₁H₁₅BrN₂O₂
MW: 287.15 g/mol
MDL Number: MFCD13189515
CAS Number: 1060813-11-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR