3-Aminoquinolin-2(1H)-one 95%

Supplier: Ambeed
Warning

A881641-100MG A881641-1G A881641-250MG
76993-744EA 18 USD
76993-744 77891-650 77891-651
3-Aminoquinolin-2(1H)-one 95%
3-Aminoquinolin-2(1H)-one

Precaución: for Research Use Only

Formula: C₉H₈N₂O
MDL Number: MFCD09754189
CAS Number: 5873-00-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR