1-Cbz-2-cyanopyrrolidine 95%

Supplier: Ambeed
Warning

A881613-1G A881613-250MG A881613-5G
77019-822EA 60.48 USD
77019-822 77019-824 77019-826
1-Cbz-2-cyanopyrrolidine 95%
1-Cbz-2-cyanopyrrolidine

Precaución: for Research Use Only

Formula: C₁₃H₁₄N₂O₂
MW: 230.26 g/mol
MDL Number: MFCD04115334
CAS Number: 119020-06-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR