4-(4-Methylbenzoyl)-1H-pyrrole-2-carboxylic acid 95%

Supplier: Ambeed
Warning

A880993-100MG A880993-1G A880993-250MG
77006-948EA 49 USD
77006-948 77006-950 77006-952
4-(4-Methylbenzoyl)-1H-pyrrole-2-carboxylic acid 95%
4-(4-Methylbenzoyl)-1H-pyrrole-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₁NO₃
MDL Number: MFCD05663824
CAS Number: 886361-19-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR