3-(p-Tolyl)propionic acid 98%

Supplier: Ambeed
Warning

A879552-100G A879552-25G A879552-5G
77006-938EA 107 USD
77006-938 77006-940 77006-942
3-(p-Tolyl)propionic acid 98%
3-(p-Tolyl)propionic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₂
MDL Number: MFCD00016561
CAS Number: 1505-50-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR