3-Chloro-6-methyl[1,2,4]triazolo[4,3-b]pyridazine 97%
Supplier: Ambeed
|
Danger
|
Synonyms:
3-Chloro-6-methyl-[1,2,4]triazolo[4,3-b]pyridazine
A879307-100MG
A879307-1G
A879307-250MG
A879307-50MG
77036-768EA
84.88
USD
77036-768
77036-770
77036-772
77036-774
3-Chloro-6-methyl[1,2,4]triazolo[4,3-b]pyridazine 97%
3-Chloro-6-methyl[1,2,4]triazolo[4,3-b]pyridazine
Precaución: for Research Use Only
Formula:
C₆H₅ClN₄ MW: 168.584 g/mol |
MDL Number:
MFCD00193097 CAS Number: 38956-68-2 UN: 2811 ADR: 6.1,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...