tert-Butyl pent-4-yn-1-ylcarbamate 95%

Supplier: Ambeed
Danger

Synonyms: N-Boc-4-pentyne-1-amine

A879100-1G A879100-250MG A879100-5G
77019-814EA 68.96 USD
77019-814 77019-816 77019-818
tert-Butyl pent-4-yn-1-ylcarbamate 95%
tert-Butyl pent-4-yn-1-ylcarbamate

Precaución: for Research Use Only

Formula: C₁₀H₁₇NO₂
MW: 183.24 g/mol
MDL Number: MFCD20036287
CAS Number: 151978-50-6
UN: 2735
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR