3-Bromoimidazo[1,2-a]pyridine-6-carbonitrile 98%

Supplier: Ambeed
Warning

Synonyms: 3-Bromo-6-cyanoimidazo[1,2-a]pyridine

A877254-1G A877254-250MG
77006-930EA 79.57 USD
77006-930 77006-932
3-Bromoimidazo[1,2-a]pyridine-6-carbonitrile 98%
3-Bromoimidazo[1,2-a]pyridine-6-carbonitrile

Precaución: for Research Use Only

Formula: C₈H₄BrN₃
MW: 222.05 g/mol
MDL Number: MFCD06659617
CAS Number: 885950-21-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR