1-Benzyl-3-phenylpiperidin-4-one 97%

Supplier: Ambeed
Warning

A877000-100MG A877000-250MG
76993-738EA 155.96 USD
76993-738 76993-740
1-Benzyl-3-phenylpiperidin-4-one 97%
1-Benzyl-3-phenylpiperidin-4-one

Precaución: for Research Use Only

Formula: C₁₈H₁₉NO
MW: 265.35 g/mol
MDL Number: MFCD11111091
CAS Number: 446302-83-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR