7-(tert-Butoxycarbonyl)-5,6,7,8-tetrahydro-1,7-naphthyridine-3-carboxylic acid 95%

Supplier: Ambeed
Warning

A874429-100MG A874429-1G A874429-250MG
77036-762EA 40.32 USD
77036-762 77036-764 77036-766
7-(tert-Butoxycarbonyl)-5,6,7,8-tetrahydro-1,7-naphthyridine-3-carboxylic acid 95%
7-(tert-Butoxycarbonyl)-5,6,7,8-tetrahydro-1,7-naphthyridine-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₈N₂O₄
MW: 278.30 g/mol
MDL Number: MFCD14706766
CAS Number: 1245645-20-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR