PIK-75 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
N'-((6-Bromoimidazo[1,2-a]pyridin-3-yl)methylene)-N,2-dimethyl-5-nitrobenzenesulphonohydrazide
A874217-10MG
A874217-1MG
A874217-25MG
A874217-5MG
76993-722EA
53.05
USD
76993-722
76993-724
76993-726
76993-728
PIK-75 95%
PIK-75
Precaución: for Research Use Only
Formula:
C₁₆H₁₄BrN₅O₄S MW: 452.3 g/mol |
MDL Number:
MFCD12407411 CAS Number: 372196-67-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...