2,2,2-Trifluoro-1-phenylethylamine hydrochloride 97%

Supplier: Ambeed
Warning

A874156-1G A874156-250MG A874156-100MG A874156-5G
77019-806EA 39 USD
77019-806 77019-808 77891-644 77891-645
2,2,2-Trifluoro-1-phenylethylamine hydrochloride 97%
2,2,2-Trifluoro-1-phenylethylamine hydrochloride

Precaución: for Research Use Only

Formula: C₈H₉ClF₃N
MDL Number: MFCD00971700
CAS Number: 13652-09-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR