4,4'-Bis(2-(9-ethyl-9H-carbazol-3-yl)vinyl)-1,1'-biphenyl 95% stabilized

Supplier: Ambeed
Warning


Stabilization: Stabilised with 4-tert-butylpyrocatechol (TBC)

A873380-100MG A873380-1G A873380-250MG
77019-800EA 92.3 USD
77019-800 77019-802 77019-804
4,4'-Bis(2-(9-ethyl-9H-carbazol-3-yl)vinyl)-1,1'-biphenyl 95% stabilized
4,4'-Bis(2-(9-ethyl-9H-carbazol-3-yl)vinyl)-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₄₄H₃₆N₂
MW: 592.77 g/mol
MDL Number: MFCD09833309
CAS Number: 475480-90-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR