1-Bromo-4,5-difluoro-2-nitrobenzene 98%

Supplier: Ambeed
Warning

A872671-10G A872671-1G A872671-5G A872671-25G A872671-100G
76993-706EA 19 USD
76993-706 76993-708 76993-710 77595-410 77891-643
1-Bromo-4,5-difluoro-2-nitrobenzene 98%
1-Bromo-4,5-difluoro-2-nitrobenzene

Precaución: for Research Use Only

Formula: C₆H₂BrF₂NO₂
MDL Number: MFCD06808551
CAS Number: 321-17-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR