2-Chloro-3-(pyrrolidin-1-ylcarbonyl)pyridine 97%

Supplier: Ambeed
Warning

Synonyms: (2-Chloropyridin-3-yl)(pyrrolidin-1-yl)methanone

A871809-1G A871809-250MG A871809-5G
77036-748EA 45.62 USD
77036-748 77036-750 77036-752
2-Chloro-3-(pyrrolidin-1-ylcarbonyl)pyridine 97%
2-Chloro-3-(pyrrolidin-1-ylcarbonyl)pyridine

Precaución: for Research Use Only

Formula: C₁₀H₁₁ClN₂O
MW: 210.66 g/mol
MDL Number: MFCD01342311
CAS Number: 60597-68-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR