1-Methylindole-4-boronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 1-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole

A871312-1G A871312-250MG A871312-5G
77036-742EA 179 USD
77036-742 77036-744 77036-746
1-Methylindole-4-boronic acid pinacol ester 98%
1-Methylindole-4-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₅H₂₀BNO₂
MW: 257.1400 g/mol
MDL Number: MFCD08690266
CAS Number: 898289-06-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR