6-Fluoro-5-methoxy-1H-indole 98%

Supplier: Ambeed
Warning

A871118-1G A871118-250MG
76993-702EA 263.11 USD
76993-702 76993-704
6-Fluoro-5-methoxy-1H-indole 98%
6-Fluoro-5-methoxy-1H-indole

Precaución: for Research Use Only

Formula: C₉H₈FNO
MW: 165.16 g/mol
MDL Number: MFCD11848568
CAS Number: 63762-83-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR