(2-Nitro-3-methoxy-phenyl)-methanol 95%

Supplier: Ambeed
Warning

Synonyms: (3-Methoxy-2-nitrophenyl)methanol

A869852-100MG A869852-1G A869852-250MG A869852-5G
76993-692EA 19 USD
76993-692 76993-694 76993-696 76993-698
(2-Nitro-3-methoxy-phenyl)-methanol 95%
(2-Nitro-3-methoxy-phenyl)-methanol

Precaución: for Research Use Only

Formula: C₈H₉NO₄
MDL Number: MFCD06202768
CAS Number: 53055-04-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR