9,10-Diphenylanthracene-2-boronic acid 95%

Supplier: Ambeed
Warning

Synonyms: (9,10-Diphenylanthracen-2-yl)boronic acid

A869338-1G A869338-250MG
77019-782EA 20 USD
77019-782 77019-784
9,10-Diphenylanthracene-2-boronic acid 95%
9,10-Diphenylanthracene-2-boronic acid

Precaución: for Research Use Only

Formula: C₂₆H₁₉BO₂
MDL Number: MFCD16621110
CAS Number: 597553-98-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR