3-(1H-Pyrazol-1-yl)aniline 97%

Supplier: Ambeed
Warning

Synonyms: 3-(1-Pyrazolyl)aniline

A869299-250MG A869299-1G
77006-924EA 50 USD
77006-924 77595-402
3-(1H-Pyrazol-1-yl)aniline 97%
3-(1H-Pyrazol-1-yl)aniline

Precaución: for Research Use Only

Formula: C₉H₉N₃
MDL Number: MFCD08059554
CAS Number: 184097-88-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR