3-Chloro-2-(difluoromethoxy)-6-(trifluoromethyl)pyridine 95%

Supplier: Ambeed
Warning

A867429-1G A867429-250MG
77036-736EA 584.56 USD
77036-736 77036-738
3-Chloro-2-(difluoromethoxy)-6-(trifluoromethyl)pyridine 95%
3-Chloro-2-(difluoromethoxy)-6-(trifluoromethyl)pyridine

Precaución: for Research Use Only

Formula: C₇H₃ClF₅NO
MW: 247.54 g/mol
CAS Number: 1214332-87-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR