3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl acetate 97%

Supplier: Ambeed
Warning

A867391-1G A867391-250MG A867391-5G
77019-764EA 114.58 USD
77019-764 77019-766 77019-768
3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl acetate 97%
3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl acetate

Precaución: for Research Use Only

Formula: C₁₅H₂₁BO₄
MW: 276.14 g/mol
MDL Number: MFCD02179491
CAS Number: 562098-07-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR