(3,5-Dimethylpyrazol-1-yl)acetic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2-(3,5-Dimethyl-1H-pyrazol-1-yl)acetic acid

A867226-25G A867226-5G
77019-760EA 284.33 USD
77019-760 77019-762
(3,5-Dimethylpyrazol-1-yl)acetic acid 97%
(3,5-Dimethylpyrazol-1-yl)acetic acid

Precaución: for Research Use Only

Formula: C₇H₁₀N₂O₂
MW: 154.17 g/mol
MDL Number: MFCD00297249
CAS Number: 16034-49-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR