3-Methoxy-4-nitropyridine 1-oxide 95%

Supplier: Ambeed
Warning

A866950-1G A866950-250MG A866950-5G
76993-680EA 148.53 USD
76993-680 76993-682 76993-684
3-Methoxy-4-nitropyridine 1-oxide 95%
3-Methoxy-4-nitropyridine 1-oxide

Precaución: for Research Use Only

Formula: C₆H₆N₂O₄
MW: 170.12 g/mol
MDL Number: MFCD16556279
CAS Number: 19355-04-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR