4-[N,N-Bis(4-bromophenyl)amino]benzaldehyde 95%, Ambeed.Inc

Supplier: Ambeed
Warning

Synonyms: 4,4'-Dibromo-4''-formyltriphenylamine, 4-(Bis(4-bromophenyl)amino)benzaldehyde

A866476-100MG A866476-250MG A866476-50MG A866476-1G
77019-754EA 55.17 USD
77019-754 77019-756 77019-758 77684-828
4-[N,N-Bis(4-bromophenyl)amino]benzaldehyde 95%, Ambeed.Inc
4-[N,N-Bis(4-bromophenyl)amino]benzaldehyde

Precaución: For research use only.

Formula: C₁₉H₁₃Br₂NO
MW: 431.13 g/mol
MDL Number: MFCD12547118
CAS Number: 25069-38-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR