2,5-Dibromo-3,4-thiophenedicarboxylic anhydride 95%

Supplier: Ambeed
Warning

Synonyms: 4,6-Dibromothieno[3,4-c]furan-1,3-dione

A866018-100MG A866018-1G A866018-250MG
77019-748EA 30 USD
77019-748 77019-750 77019-752
2,5-Dibromo-3,4-thiophenedicarboxylic anhydride 95%
2,5-Dibromo-3,4-thiophenedicarboxylic anhydride

Precaución: for Research Use Only

Formula: C₆Br₂O₃S
MW: 311.93 g/mol
MDL Number: MFCD27923031
CAS Number: 1015423-45-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR