2-Chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3-(trifluoromethyl)pyridine 98%

Supplier: Ambeed
Warning

A865311-100MG A865311-250MG A865311-1G
77036-732EA 19 USD
77036-732 77036-734 77873-326
2-Chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3-(trifluoromethyl)pyridine 98%
2-Chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3-(trifluoromethyl)pyridine

Precaución: for Research Use Only

Formula: C₁₂H₁₄BClF₃NO₂
MDL Number: MFCD12923433
CAS Number: 741709-67-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR