2-Aminobiphenyl Hydrochloride 97%

Supplier: Ambeed
Warning

A864651-1G A864651-5G
77019-746EA 12 USD
77019-746 77573-744
2-Aminobiphenyl Hydrochloride 97%
2-Aminobiphenyl Hydrochloride

Precaución: for Research Use Only

Formula: C₁₂H₁₂ClN
MDL Number: MFCD00035473
CAS Number: 2185-92-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR