Methyl-7-chlorobenzo[b]thiophene-2-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Methyl 7-chlorobenzo[b]thiophene-2-carboxylate

A864193-100MG A864193-1G A864193-250MG A864193-5G
77006-906EA 13.8 USD
77006-906 77006-908 77006-910 77674-284
Methyl-7-chlorobenzo[b]thiophene-2-carboxylate 95%
Methyl 7-chlorobenzo[b]thiophene-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₇ClO₂S
MW: 226.6794 g/mol
MDL Number: MFCD09027105
CAS Number: 550998-56-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR