Methyl 5-fluoro-3-formyl-1H-indole-2-carboxylate 95%

Supplier: Ambeed
Warning

A863568-100MG A863568-1G A863568-250MG
77019-740EA 68.96 USD
77019-740 77019-742 77019-744
Methyl 5-fluoro-3-formyl-1H-indole-2-carboxylate 95%
Methyl 5-fluoro-3-formyl-1H-indole-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₈FNO₃
MW: 221.18 g/mol
MDL Number: MFCD03848199
CAS Number: 843629-51-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR