2-(2-Thienyl)benzothiazole 98%

Supplier: Ambeed
Warning

A863029-1G A863029-25G A863029-5G
77019-734EA 32 USD
77019-734 77019-736 77019-738
2-(2-Thienyl)benzothiazole 98%
2-(2-Thienyl)benzothiazole

Precaución: for Research Use Only

Formula: C₁₁H₇NS₂
MW: 217.30 g/mol
MDL Number: MFCD00179918
CAS Number: 34243-38-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR