1-(Benzo[d][1,3]dioxol-5-yl)ethanol 95%

Supplier: Ambeed
Warning

A862981-1G A862981-250MG
76993-664EA 222.79 USD
76993-664 76993-666
1-(Benzo[d][1,3]dioxol-5-yl)ethanol 95%
1-(Benzo[d][1,3]dioxol-5-yl)ethanol

Precaución: for Research Use Only

Formula: C₉H₁₀O₃
MW: 166.17 g/mol
MDL Number: MFCD11149380
CAS Number: 6329-73-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR