6-Bromo-2-fluoro-3-methoxypyridine 95%

Supplier: Ambeed
Warning

A862613-100MG A862613-1G A862613-250MG
77036-720EA 15 USD
77036-720 77036-722 77036-724
6-Bromo-2-fluoro-3-methoxypyridine 95%
6-Bromo-2-fluoro-3-methoxypyridine

Precaución: for Research Use Only

Formula: C₆H₅BrFNO
MDL Number: MFCD04112568
CAS Number: 850142-73-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR