(S)-N-Fmoc-azetidine-2-carboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: (S)-1-(((9H-Fluoren-9-yl)methoxy)carbonyl)azetidine-2-carboxylic acid, Fmoc-L-azetidine-2-carboxylic acid

A862109-10G A862109-1G A862109-250MG A862109-5G
77036-712EA 140 USD
77036-712 77036-714 77036-716 77036-718
(S)-N-Fmoc-azetidine-2-carboxylic acid 97%
(S)-N-Fmoc-azetidine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₉H₁₇NO₄
MDL Number: MFCD01320843
CAS Number: 136552-06-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR