Octadecyl 2,2,2-trifluoroacetate 95%

Supplier: Ambeed
Warning

A861707-100MG A861707-10G A861707-1G A861707-250MG A861707-5G
77019-720EA 15 USD
77019-720 77019-722 77019-724 77019-726 77019-728
Octadecyl 2,2,2-trifluoroacetate 95%
Octadecyl 2,2,2-trifluoroacetate

Precaución: for Research Use Only

Formula: C₂₀H₃₇F₃O₂
MW: 366.50 g/mol
MDL Number: MFCD26792329
CAS Number: 79392-43-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR