4-Bromo-2,6-dimethylbenzenecarbonitrile 95%

Supplier: Ambeed
Warning

A860825-1G A860825-250MG A860825-5G
77006-888EA 78 USD
77006-888 77006-890 77882-786
4-Bromo-2,6-dimethylbenzenecarbonitrile 95%
4-Bromo-2,6-dimethylbenzenecarbonitrile

Precaución: for Research Use Only

Formula: C₉H₈BrN
MDL Number: MFCD01463960
CAS Number: 5757-66-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR