3-Fluoro-2-hydroxy-5-(trifluoromethyl)pyridine 98%

Supplier: Ambeed
Warning

Synonyms: 3-Fluoro-5-(trifluoromethyl)pyridin-2-ol

A860487-100MG A860487-250MG
77036-708EA 42 USD
77036-708 77036-710
3-Fluoro-2-hydroxy-5-(trifluoromethyl)pyridine 98%
3-Fluoro-2-hydroxy-5-(trifluoromethyl)pyridine

Precaución: for Research Use Only

Formula: C₆H₃F₄NO
MDL Number: MFCD10699119
CAS Number: 1040683-15-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR