Ethyl thiophene-2-carboxylate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Ethyl 2-thiophenecarboxylate
A859951-100G
A859951-25G
A859951-5G
A859951-500G
77006-882EA
77
USD
77006-882
77006-884
77006-886
77674-270
Ethyl thiophene-2-carboxylate 98%
Ethyl thiophene-2-carboxylate
Precaución: for Research Use Only
Formula:
C₇H₈O₂S |
MDL Number:
MFCD00005436 CAS Number: 2810-04-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...