Methyl 2-aminobenzo[d]thiazole-4-carboxylate 97%

Supplier: Ambeed
Warning

A859656-1G A859656-250MG
76993-642EA 234.46 USD
76993-642 77684-819
Methyl 2-aminobenzo[d]thiazole-4-carboxylate 97%
Methyl 2-aminobenzo[d]thiazole-4-carboxylate

Precaución: For research use only.

Formula: C₉H₈N₂O₂S
MW: 208.23 g/mol
MDL Number: MFCD04971761
CAS Number: 1024054-68-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR