6-Bromobenzo[b]thiophene-3-carbaldehyde ≥95%

Supplier: Ambeed
Warning

A859376-100MG A859376-250MG
77019-716EA 238 USD
77019-716 77019-718
6-Bromobenzo[b]thiophene-3-carbaldehyde ≥95%
6-Bromobenzo[b]thiophene-3-carbaldehyde

Precaución: for Research Use Only

Formula: C₉H₅BrOS
MDL Number: MFCD25542199
CAS Number: 19075-53-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR