tert-Butyl 6-amino-7-fluoro-3,4-dihydroisoquinoline-2(1H)-carboxylate 95%

Supplier: Ambeed
Warning

A857782-100MG A857782-1G A857782-250MG
76993-636EA 51.99 USD
76993-636 76993-638 76993-640
tert-Butyl 6-amino-7-fluoro-3,4-dihydroisoquinoline-2(1H)-carboxylate 95%
tert-Butyl 6-amino-7-fluoro-3,4-dihydroisoquinoline-2(1H)-carboxylate

Precaución: for Research Use Only

Formula: C₁₄H₁₉FN₂O₂
MW: 266.31 g/mol
MDL Number: MFCD18157611
CAS Number: 912846-68-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR