(1R,2S,5R)-2-Isopropyl-5-methylcyclohexyl-2,2-dihydroxyacetate 98%

Supplier: Ambeed
Danger

A857378-10G A857378-1G A857378-5G A857378-25G
77006-868EA 15 USD
77006-868 77006-870 77674-268 77882-780
(1R,2S,5R)-2-Isopropyl-5-methylcyclohexyl-2,2-dihydroxyacetate 98%
(1R,2S,5R)-2-Isopropyl-5-methylcyclohexyl-2,2-dihydroxyacetate

Precaución: for Research Use Only

Formula: C₁₂H₂₂O₄
MDL Number: MFCD00792503
CAS Number: 111969-64-3
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR