Hexahydropyrrolo[1,2-a]pyrazin-6(2H)-one 97%

Supplier: Ambeed
Danger

A857368-1G A857368-5G A857368-250MG
77019-712EA 272.66 USD
77019-712 77019-714 77614-528
Hexahydropyrrolo[1,2-a]pyrazin-6(2H)-one 97%
Hexahydropyrrolo[1,2-a]pyrazin-6(2H)-one

Precaución: for Research Use Only

Formula: C₇H₁₂N₂O
MW: 140.18 g/mol
MDL Number: MFCD08752614
CAS Number: 117810-52-3
UN: 3259
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR