Z-Dehydro-Ala-OMe 98%

Supplier: Ambeed
Warning

A856387-1G A856387-250MG A856387-5G
77006-862EA 120 USD
77006-862 77006-864 77882-779
Z-Dehydro-Ala-OMe 98%
Z-Dehydro-Ala-OMe

Precaución: for Research Use Only

Formula: C₁₂H₁₃NO₄
MDL Number: MFCD00056245
CAS Number: 21149-17-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR