4,4'-Diphenoxydiphenylsulfone 97%

Supplier: Ambeed
Warning

Synonyms: 4,4'-sulphonylbis(phenoxybenzene)

A855594-100MG A855594-1G A855594-250MG
76993-624EA 14 USD
76993-624 76993-626 76993-628
4,4'-Diphenoxydiphenylsulfone 97%
4,4'-Diphenoxydiphenylsulfone

Precaución: for Research Use Only

Formula: C₂₄H₁₈O₄S
MW: 402.462 g/mol
MDL Number: MFCD00221991
CAS Number: 1623-91-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR