Methyl-5-bromo-2-(bromomethyl)benzoate 95%
Supplier: Ambeed
|
Danger
|
A855519-250MG
A855519-25G
A855519-1G
A855519-5G
A855519-100MG
77036-706EA
18
USD
77036-706
77573-716
77670-238
77670-240
77873-318
Methyl-5-bromo-2-(bromomethyl)benzoate 95%
Methyl-5-bromo-2-(bromomethyl)benzoate
Precaución: for Research Use Only
Formula:
C₉H₈Br₂O₂ |
MDL Number:
MFCD11505949 CAS Number: 79670-17-0 UN: 3261 ADR: 8,Ⅱ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...