(2R,5S)-rel-Diethyl 2,5-dibromohexanedioate 98%

Supplier: Ambeed
Warning

Synonyms: (2R,5S)-rel-Diethyl-2,5-dibromoadipate, (2R,5S)-rel-2,5-Dibromoadipic acid diethyl ester

A855502-100G A855502-25G A855502-500G
77036-702EA 74.27 USD
77036-702 77036-704 77670-236
(2R,5S)-rel-Diethyl 2,5-dibromohexanedioate 98%
(2R,5S)-rel-Diethyl 2,5-dibromohexanedioate

Precaución: for Research Use Only

Formula: C₁₀H₁₆Br₂O₄
MW: 360.04 g/mol
MDL Number: MFCD00075379
CAS Number: 54221-37-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR