(R)-(+)-2-Methyl-CBS-oxazaborolidine monohydrate 97%

Supplier: Ambeed
Warning

Synonyms: (R)-2-Methyl-CBS-oxazaborolidine monohydrate, (R)-1-Methyl-3,3-diphenylhexahydropyrrolo[1,2-c][1,3,2]oxazaborole, (R)-3,3-Diphenyl-1-methylpyrrolidino[1,2-c]-1,3,2-oxazaborole, α,α-Diphenyl-D-prolinolmethylboronic acid cyclamide ester

A855211-100G A855211-1G A855211-25G A855211-500G A855211-5G
77006-852EA 176.11 USD
77006-852 77006-854 77006-856 77006-858 77006-860
(R)-(+)-2-Methyl-CBS-oxazaborolidine monohydrate 97%
(R)-(+)-2-Methyl-CBS-oxazaborolidine monohydrate

Precaución: for Research Use Only

Formula: C₁₈H₂₀BNO·1H₂O
MW: 295.19 g/mol (277.17 anhy)
MDL Number: MFCD00078440
CAS Number: 112022-83-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR